Search
Search

ERLIN2| Gene Symbol: ERLIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424380.3 | XP_424380.1 | 5 - 6 | 485 | 67 | |
| GenBank mRNA | BX930312.1 | - | 5 - 6 | 460 | 67 | |
| BX932498.1 | - | 5 - 6 | 452 | 67 | ||
Set Membership: |
|