Search
Search

CREB5| Gene Symbol: CREB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281825.2 | XP_015137311.1 | 3 - 4 | 284 | 72 | |
| XM_025147446.1 | XP_025003214.1 | 4 - 5 | 364 | 72 | ||
| XM_425995.6 | XP_425995.3 | 4 - 5 | 364 | 72 | ||
| GenBank mRNA | BX930883.1 | - | 4 - 5 | 330 | 72 | |
| BX933040.2 | - | 4 - 5 | 337 | 72 | ||
Set Membership: |
|