Search
Search

TSR3| Gene Symbol: TSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277169.1 | NP_001264098.1 | 3 - 4 | 563 | 87 | |
| GenBank mRNA | BX930961.1 | - | 3 - 4 | 543 | 87 | |
| BX931234.1 | - | 3 - 4 | 532 | 87 | ||
| BX932485.1 | - | 3 - 4 | 523 | 87 | ||
Set Membership: |
|