Search
Search

RBP7| Gene Symbol: RBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417606.5 | XP_417606.4 | 2 - 3 | 304 | 121 | |
| GenBank mRNA | BX929289.2 | - | 2 - 3 | 269 | 121 | |
| BX931240.1 | - | 2 - 3 | 274 | 121 | ||
| BX934511.1 | - | 2 - 3 | 319 | 121 | ||
| BX934521.1 | - | 2 - 3 | 287 | 121 | ||
Set Membership: |
|