Search
Search

CTSV| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168009.1 | NP_001161481.1 | 1 - 2 | 53 | 58 | |
| GenBank mRNA | BX932251.2 | - | 1 - 2 | 39 | 58 | |
| BX933970.1 | - | 1 - 2 | 40 | 58 | ||
| BX936191.1 | - | 1 - 2 | 27 | 58 | ||
Set Membership: |
|