Search
Search

IDS| Gene Symbol: IDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277660.1 | NP_001264589.1 | 4 - 5 | 519 | 88 | |
| XM_015278303.2 | XP_015133789.2 | 5 - 6 | 551 | 88 | ||
| XM_015278304.2 | XP_015133790.1 | 5 - 6 | 667 | 88 | ||
| XM_015278305.2 | XP_015133791.1 | 5 - 6 | 654 | 88 | ||
| XM_015278306.2 | XP_015133792.1 | 4 - 5 | 585 | 88 | ||
| XM_015278307.2 | XP_015133793.1 | 5 - 6 | 633 | 88 | ||
| XM_015278308.2 | XP_015133794.1 | 5 - 6 | 535 | 88 | ||
| XM_025149575.1 | XP_025005343.1 | 5 - 6 | 624 | 88 | ||
| XM_025149576.1 | XP_025005344.1 | 6 - 7 | 623 | 88 | ||
| XM_025149577.1 | XP_025005345.1 | 6 - 7 | 638 | 88 | ||
| XM_025149578.1 | XP_025005346.1 | 4 - 5 | 525 | 88 | ||
| GenBank mRNA | BU247591.1 | - | 4 - 5 | 510 | 88 | |
| BX934826.1 | - | 4 - 5 | 511 | 88 | ||
| CD215596.1 | - | 4 - 5 | 515 | 88 | ||
Set Membership: |
|