Search
Search

DNAAF5| Gene Symbol: DNAAF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414755.5 | XP_414755.2 | 7 - 8 | 1588 | 92 | |
| GenBank mRNA | BX934565.2 | - | 5 - 6 | 750 | 92 | |
| BX936040.1 | - | 5 - 6 | 758 | 92 | ||
Set Membership: |
|