Search
Search

DCP1A| Gene Symbol: DCP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944566.3 | XP_004944623.1 | 5 - 6 | 553 | 93 | |
| XM_004944567.3 | XP_004944624.1 | 5 - 6 | 553 | 93 | ||
| XM_015293406.2 | XP_015148892.1 | 5 - 6 | 551 | 93 | ||
| XM_025154556.1 | XP_025010324.1 | 5 - 6 | 553 | 93 | ||
| GenBank mRNA | CR353352.1 | - | 5 - 6 | 522 | 93 | |
Set Membership: |
|