Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Gg03322359_m1
          See other RALGAPA1 GE Assays ›
          Gene Symbol:
          RALGAPA1
          Gene Name:
          Ral GTPase activating protein catalytic alpha subunit 1
          Gene Aliases:
          GARNL1
          Chromosome Location:
          Chr.5: 36443905 - 36556629 on Build GRCg6a
          Species:
          Chicken
          Species Specific ID (Flybase ID):
          -
          Assay ID Gg03322359_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RALGAPA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277447.2 NP_001264376.1 11 - 12 1661 70
          NM_001277451.2 NP_001264380.1 11 - 12 1661 70
          NM_001277452.2 NP_001264381.1 11 - 12 1661 70
          NM_001277453.2 NP_001264382.1 11 - 12 1661 70
          NM_001277454.2 NP_001264383.1 11 - 12 1661 70
          XM_015287370.2 XP_015142856.1 11 - 12 1713 70
          XM_015287371.2 XP_015142857.1 11 - 12 1712 70
          XM_015287372.2 XP_015142858.1 11 - 12 1712 70
          XM_015287373.2 XP_015142859.1 11 - 12 1712 70
          XM_015287374.2 XP_015142860.1 11 - 12 1713 70
          XM_015287375.2 XP_015142861.1 11 - 12 1712 70
          XM_015287376.2 XP_015142862.1 11 - 12 1712 70
          XM_015287377.2 XP_015142863.1 11 - 12 1713 70
          XM_015287378.2 XP_015142864.1 11 - 12 1710 70
          XM_015287379.2 XP_015142865.1 11 - 12 1709 70
          XM_015287380.2 XP_015142866.1 11 - 12 1445 70
          XM_015287381.2 XP_015142867.1 11 - 12 1445 70
          XM_015287383.2 XP_015142869.1 11 - 12 1445 70
          XM_025150740.1 XP_025006508.1 11 - 12 1713 70
          XM_025150741.1 XP_025006509.1 11 - 12 1713 70
          XM_025150742.1 XP_025006510.1 11 - 12 1445 70
          GenBank mRNA BX929992.2 - 6 - 7 996 70
          BX933468.2 - 1 - 2 200 70
          JF330255.1 - 11 - 12 1568 70
          JF330256.1 - 11 - 12 1568 70
          JF330257.1 - 11 - 12 1568 70
          JF330258.1 - 11 - 12 1568 70
          JF330259.1 - 11 - 12 1568 70

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline