Search
Search

TAF7| Gene Symbol: TAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277886.1 | NP_001264815.1 | 5 - 6 | 505 | 93 | |
| XM_015278223.2 | XP_015133709.1 | 5 - 6 | 520 | 93 | ||
| XM_015278224.2 | XP_015133710.1 | 5 - 6 | 521 | 93 | ||
| XM_025149547.1 | XP_025005315.1 | 5 - 6 | 521 | 93 | ||
| GenBank mRNA | BU112689.1 | - | 5 - 6 | 506 | 93 | |
| BX930950.1 | - | 5 - 6 | 491 | 93 | ||
| BX935900.2 | - | 5 - 6 | 507 | 93 | ||
Set Membership: |
|