Search
Search

SYT12| Gene Symbol: SYT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199512.1 | NP_001186441.1 | 5 - 6 | 926 | 64 | |
| GenBank mRNA | BU365478.1 | - | 2 - 3 | 297 | 64 | |
| BX936066.2 | - | 4 - 5 | 398 | 64 | ||
Set Membership: |
|