Search
Search

DAP3| Gene Symbol: DAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298658.2 | XP_015154144.1 | 5 - 6 | 544 | 59 | |
| XM_025143521.1 | XP_024999289.1 | 5 - 6 | 459 | 59 | ||
| XM_422859.6 | XP_422859.3 | 5 - 6 | 447 | 59 | ||
| GenBank mRNA | BX929536.1 | - | 5 - 6 | 415 | 59 | |
Set Membership: |
|