Search
Search

LONRF3| Gene Symbol: LONRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940846.2 | XP_004940903.1 | 5 - 6 | 1328 | 75 | |
| XM_025149880.1 | XP_025005648.1 | 4 - 5 | 1172 | 75 | ||
| XM_025149881.1 | XP_025005649.1 | 3 - 4 | 1049 | 75 | ||
| GenBank mRNA | CR389285.1 | - | 1 - 2 | 238 | 75 | |
Set Membership: |
|