Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Gg03324558_m1
          See other ABI3BP GE Assays ›
          Gene Symbol:
          ABI3BP
          Gene Name:
          ABI family member 3 binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 86110406 - 86243541 on Build GRCg6a
          Species:
          Chicken
          Species Specific ID (Flybase ID):
          -
          Assay ID Gg03324558_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ABI3BP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_015296923.2 XP_015152409.1 34 - 35 2761 75
          XM_015296932.2 XP_015152418.1 34 - 35 2761 75
          XM_015296937.2 XP_015152423.1 34 - 35 2749 75
          XM_015296941.2 XP_015152427.1 33 - 34 2713 75
          XM_015296943.2 XP_015152429.1 33 - 34 2713 75
          XM_015296948.2 XP_015152434.1 33 - 34 2695 75
          XM_015296952.2 XP_015152438.1 33 - 34 2695 75
          XM_015296959.2 XP_015152445.1 33 - 34 2695 75
          XM_015296967.2 XP_015152453.1 33 - 34 2686 75
          XM_015296978.2 XP_015152464.1 33 - 34 2686 75
          XM_015296987.2 XP_015152473.1 33 - 34 2683 75
          XM_015296995.2 XP_015152481.1 33 - 34 2683 75
          XM_015297002.2 XP_015152488.1 32 - 33 2665 75
          XM_015297007.1 XP_015152493.1 34 - 35 2762 75
          XM_015297017.1 XP_015152503.1 34 - 35 2762 75
          XM_015297025.2 XP_015152511.1 31 - 32 2590 75
          XM_015297032.2 XP_015152518.1 30 - 31 2490 75
          XM_015297034.1 XP_015152520.1 34 - 35 2762 75
          XM_015297037.2 XP_015152523.1 28 - 29 2339 75
          XM_015297043.2 XP_015152529.1 27 - 28 2291 75
          XM_015297048.2 XP_015152534.1 27 - 28 2274 75
          XM_015297057.2 XP_015152543.1 27 - 28 2259 75
          XM_015297064.2 XP_015152550.1 26 - 27 2216 75
          XM_015297073.2 XP_015152559.1 26 - 27 2192 75
          XM_015297083.2 XP_015152569.1 26 - 27 2180 75
          XM_015297088.2 XP_015152574.1 25 - 26 2114 75
          XM_015297091.2 XP_015152577.1 24 - 25 2084 75
          XM_025143975.1 XP_024999743.1 32 - 33 2647 75
          XM_025143979.1 XP_024999747.1 32 - 33 2608 75
          XM_025143986.1 XP_024999754.1 29 - 30 2442 75
          XM_025144007.1 XP_024999775.1 24 - 25 2039 75
          GenBank mRNA CR389415.1 - 9 - 10 617 75
          CR390813.1 - 10 - 11 646 75

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline