Search
Search

ANGPTL5| Gene Symbol: ANGPTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197236.1 | NP_001184165.1 | 2 - 3 | 380 | 148 | |
| GenBank mRNA | CR389624.1 | - | 2 - 3 | 380 | 148 | |
Set Membership: |
|