Search
Search

RPS6KC1| Gene Symbol: RPS6KC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935322.3 | XP_004935379.1 | 10 - 11 | 1228 | 70 | |
| XM_004935324.3 | XP_004935381.1 | 10 - 11 | 1229 | 70 | ||
| XM_015283890.2 | XP_015139376.1 | 9 - 10 | 1192 | 70 | ||
| XM_015283891.2 | XP_015139377.1 | 11 - 12 | 1273 | 70 | ||
| XM_025148969.1 | XP_025004737.1 | 9 - 10 | 1136 | 70 | ||
| GenBank mRNA | CR524437.1 | - | 4 - 5 | 544 | 70 | |
Set Membership: |
|