Search
Search

ABCC2| Gene Symbol: ABCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288821.2 | XP_015144307.1 | 31 - 32 | 4952 | 60 | |
| XM_025151804.1 | XP_025007572.1 | 30 - 31 | 4817 | 60 | ||
| GenBank mRNA | CR733321.1 | - | 6 - 7 | 789 | 60 | |
Set Membership: |
|