Search
Search

SCP2| Gene Symbol: SCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305200.1 | NP_001292129.1 | 3 - 4 | 217 | 60 | |
| GenBank mRNA | BU114627.1 | - | 220 | 60 | ||
| CR389316.1 | - | 216 | 60 | |||
| L09231.1 | - | 3 - 4 | 212 | 60 | ||
Set Membership: |
|