Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: TMEM45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278043.1 | NP_001264972.1 | 2 - 3 | 249 | 65 | |
XM_015296096.2 | XP_015151582.1 | 2 - 3 | 314 | 65 | ||
XM_015296102.2 | XP_015151588.1 | 2 - 3 | 506 | 65 | ||
XM_015296108.2 | XP_015151594.1 | 2 - 3 | 456 | 65 | ||
GenBank mRNA | BX931403.2 | - | 2 - 3 | 249 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |