Search
Search

TSPO2| Gene Symbol: TSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004948410.3 | XP_004948467.1 | 437 | 62 | ||
| XM_418037.5 | XP_418037.2 | 487 | 62 | |||
| GenBank mRNA | BX931454.2 | - | 428 | 62 | ||
| BX934347.2 | - | 222 | 62 | |||
| BX936041.2 | - | 419 | 62 | |||
| CR353324.1 | - | 428 | 62 | |||
Set Membership: |
|