Search
Search

FBXO30| Gene Symbol: FBXO30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284302.2 | XP_015139788.1 | 2 - 3 | 311 | 69 | |
| XM_015284303.2 | XP_015139789.1 | 2 - 3 | 147 | 69 | ||
| XM_025149424.1 | XP_025005192.1 | 1 - 2 | 120 | 69 | ||
| GenBank mRNA | BX932343.2 | - | 1 - 2 | 263 | 69 | |
Set Membership: |
|