Search
Search

PCSK5| Gene Symbol: PCSK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280354.2 | XP_015135840.1 | 3 - 4 | 865 | 63 | |
| XM_424841.6 | XP_424841.4 | 3 - 4 | 865 | 63 | ||
| GenBank mRNA | CR352595.1 | - | 3 - 4 | 432 | 63 | |
Set Membership: |
|