Search
Search

CYP3A4| Gene Symbol: CYP3A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329508.1 | NP_001316437.1 | 4 - 5 | 357 | 73 | |
| XM_015294357.2 | XP_015149843.1 | 4 - 5 | 408 | 73 | ||
| GenBank mRNA | BX931257.1 | - | 4 - 5 | 358 | 73 | |
Set Membership: |
|