Search
Search

SUGP2| Gene Symbol: SUGP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195569.1 | NP_001182498.1 | 1 - 2 | 348 | 146 | |
| XM_015300047.2 | XP_015155533.1 | 1 - 2 | 343 | 146 | ||
| XM_015300048.2 | XP_015155534.1 | 1 - 2 | 343 | 146 | ||
| XM_015300049.2 | XP_015155535.1 | 1 - 2 | 343 | 146 | ||
| XM_015300050.2 | XP_015155536.1 | 1 - 2 | 343 | 146 | ||
| XM_025144274.1 | XP_025000042.1 | 2 - 3 | 222 | 146 | ||
| XM_025144275.1 | XP_025000043.1 | 2 - 3 | 560 | 146 | ||
| GenBank mRNA | CR391045.1 | - | 1 - 2 | 214 | 146 | |
Set Membership: |
|