Search
Search

METTL2B| Gene Symbol: METTL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006329.1 | NP_001006329.1 | 1 - 2 | 117 | 56 | |
| GenBank mRNA | AJ721086.1 | - | 1 - 2 | 117 | 56 | |
| BX931317.2 | - | 1 - 2 | 77 | 56 | ||
Set Membership: |
|