Search
Search

SSBP3| Gene Symbol: SSBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204564.1 | NP_989895.1 | 5 - 6 | 379 | 51 | |
| XM_015290944.2 | XP_015146430.1 | 5 - 6 | 538 | 51 | ||
| GenBank mRNA | CR386174.1 | - | 5 - 6 | 544 | 51 | |
| U68380.2 | - | 5 - 6 | 379 | 51 | ||
Set Membership: |
|