Search
Search

BOK| Gene Symbol: BOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204706.1 | NP_990037.1 | 4 - 5 | 757 | 124 | |
| GenBank mRNA | AF275944.1 | - | 4 - 5 | 757 | 124 | |
| AF290888.1 | - | 3 - 4 | 568 | 124 | ||
Set Membership: |
|