Search
Search

ATP6V0A1| Gene Symbol: ATP6V0A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204724.2 | NP_990055.1 | 14 - 15 | 1616 | 63 | |
| XM_015299582.2 | XP_015155068.1 | 14 - 15 | 1684 | 63 | ||
| XM_015299583.2 | XP_015155069.1 | 14 - 15 | 1684 | 63 | ||
| XM_015299584.2 | XP_015155070.1 | 14 - 15 | 1663 | 63 | ||
| XM_015299585.2 | XP_015155071.1 | 14 - 15 | 1684 | 63 | ||
| XM_015299586.2 | XP_015155072.1 | 14 - 15 | 1684 | 63 | ||
| XM_015299587.2 | XP_015155073.1 | 14 - 15 | 1663 | 63 | ||
| GenBank mRNA | AJ289019.1 | - | 14 - 15 | 1621 | 63 | |
Set Membership: |
|