Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Gg03339394_m1
          See other TNNT3 GE Assays ›
          Gene Symbol:
          TNNT3
          Gene Name:
          troponin T3, fast skeletal type
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 14207179 - 14241384 on Build GRCg6a
          Species:
          Chicken
          Species Specific ID (Flybase ID):
          -
          Assay ID Gg03339394_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNNT3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_204922.2 NP_990253.1 20 - 21 738 74
          XM_015286531.2 XP_015142017.1 23 - 24 821 74
          XM_015286532.1 XP_015142018.1 22 - 23 809 74
          XM_015286533.1 XP_015142019.1 22 - 23 809 74
          XM_015286534.2 XP_015142020.1 22 - 23 806 74
          XM_015286535.1 XP_015142021.1 22 - 23 803 74
          XM_015286537.1 XP_015142023.1 21 - 22 797 74
          XM_015286538.1 XP_015142024.1 21 - 22 792 74
          XM_015286539.1 XP_015142025.1 21 - 22 788 74
          XM_015286540.2 XP_015142026.1 20 - 21 779 74
          XM_015286541.1 XP_015142027.1 20 - 21 777 74
          XM_015286542.2 XP_015142028.1 20 - 21 773 74
          XM_015286543.2 XP_015142029.1 20 - 21 773 74
          XM_015286544.2 XP_015142030.1 19 - 20 767 74
          XM_015286545.2 XP_015142031.1 19 - 20 764 74
          XM_015286546.2 XP_015142032.1 19 - 20 764 74
          XM_015286548.2 XP_015142034.1 19 - 20 761 74
          XM_015286549.2 XP_015142035.1 19 - 20 761 74
          XM_015286550.2 XP_015142036.1 19 - 20 761 74
          XM_015286551.2 XP_015142037.1 19 - 20 758 74
          XM_015286552.2 XP_015142038.1 19 - 20 758 74
          XM_015286553.2 XP_015142039.1 18 - 19 752 74
          XM_015286554.2 XP_015142040.1 18 - 19 749 74
          XM_015286555.2 XP_015142041.1 18 - 19 749 74
          XM_015286556.2 XP_015142042.1 18 - 19 746 74
          XM_015286557.2 XP_015142043.1 18 - 19 743 74
          XM_015286559.2 XP_015142045.1 17 - 18 737 74
          XM_015286560.2 XP_015142046.1 17 - 18 734 74
          XM_015286561.2 XP_015142047.1 17 - 18 734 74
          XM_015286562.2 XP_015142048.1 17 - 18 734 74
          XM_015286563.1 XP_015142049.1 17 - 18 728 74
          XM_015286564.2 XP_015142050.1 16 - 17 722 74
          XM_015286565.1 XP_015142051.1 16 - 17 713 74
          XM_015286566.1 XP_015142052.1 15 - 16 704 74
          XM_015286567.1 XP_015142053.1 15 - 16 701 74
          XM_015286568.1 XP_015142054.1 15 - 16 701 74
          XM_015286570.2 XP_015142056.1 15 - 16 701 74
          XM_015286571.1 XP_015142057.1 15 - 16 695 74
          XM_015286572.1 XP_015142058.1 14 - 15 689 74
          XM_015286573.1 XP_015142059.1 14 - 15 683 74
          XM_015286574.1 XP_015142060.1 14 - 15 683 74
          XM_015286575.2 XP_015142061.1 13 - 14 670 74
          XM_015286576.1 XP_015142062.1 13 - 14 665 74
          GenBank mRNA AF044922.1 - 20 - 21 739 74
          BX931434.2 - 15 - 16 720 74
          M22154.1 - 17 - 18 695 74
          M22155.1 - 14 - 15 649 74
          M22156.1 - 15 - 16 664 74
          M22158.1 - 13 - 14 628 74

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          muscle contraction
          regulation of striated muscle contraction
          sarcomere organization
          tropomyosin binding
          troponin C binding
          troponin I binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline