Search
Search

KBP| Gene Symbol: KBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205353.2 | NP_990684.1 | 4 - 5 | 288 | 87 | |
| XM_025142488.1 | XP_024998256.1 | 4 - 5 | 1109 | 87 | ||
| GenBank mRNA | BI394598.1 | - | 4 - 5 | 288 | 87 | |
| X17700.1 | - | 280 | 87 | |||
Set Membership: |
|