Search
Search

TGIF1| Gene Symbol: TGIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205379.1 | NP_990710.1 | 2 - 3 | 491 | 67 | |
| XM_015282585.2 | XP_015138071.1 | 2 - 3 | 736 | 67 | ||
| XM_025147068.1 | XP_025002836.1 | 2 - 3 | 547 | 67 | ||
| GenBank mRNA | U25353.1 | - | 2 - 3 | 491 | 67 | |
Set Membership: |
|