Search
Search

RBP| Gene Symbol: RBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205463.2 | NP_990794.1 | 4 - 5 | 428 | 134 | |
| XM_025152771.1 | XP_025008539.1 | 4 - 5 | 3133 | 134 | ||
| GenBank mRNA | BU267624.1 | - | 4 - 5 | 438 | 134 | |
| CR353084.1 | - | 4 - 5 | 428 | 134 | ||
| J03922.1 | - | 4 - 5 | 433 | 134 | ||
| X74247.1 | - | 409 | 134 | |||
Set Membership: |
|