Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Gg03340091_m1
See other PCK1 GE Assays ›
Gene Symbol:
PCK1
Gene Name:
phosphoenolpyruvate carboxykinase 1
Gene Aliases:
PEPCK
Chromosome Location:
Chr.20: 11554555 - 11562277 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID
Gg03340091_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PCK1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_205471.1
NP_990802.1
8 - 9
1569
85
GenBank mRNA
M14229.1
-
8 - 9
1569
85
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Gallus gallus
Probe spans exons
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
lactate metabolic process
pyruvate metabolic process
gluconeogenesis
regulation of gluconeogenesis
nucleoside diphosphate phosphorylation
alanine metabolic process
aspartate metabolic process
glutamate metabolic process
glutamine metabolic process
glycine metabolic process
proline metabolic process
NADH regeneration
vitellogenesis
aging
digestion
adult feeding behavior
serine family amino acid metabolic process
embryo development ending in birth or egg hatching
bicarbonate transport
malate transport
lipid catabolic process
peptidyl-serine phosphorylation
naphthalenesulfonate metabolic process
oviposition
propionate catabolic process
glycerol catabolic process
sexual reproduction
eggshell formation
response to lipopolysaccharide
cellular response to insulin stimulus
response to lipid
cellular response to glucose starvation
response to starvation
positive regulation of memory T cell differentiation
positive regulation of cAMP-mediated signaling
positive regulation of gluconeogenesis
negative regulation of carbohydrate metabolic process
positive regulation of carbohydrate metabolic process
regulation of transcription by glucose
regulation of lipid biosynthetic process
response to cycloheximide
embryonic organ morphogenesis
developmental growth
intestinal absorption
response to glucocorticoid
response to cAMP
regulation of feeding behavior
hepatocyte differentiation
response to interleukin-6
cellular response to retinoic acid
cellular response to cAMP
cellular response to fructose stimulus
cellular response to glucose stimulus
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to ethanol
cellular response to parathyroid hormone stimulus
cellular response to glucagon stimulus
cellular response to hypoxia
cellular response to dexamethasone stimulus
kidney interstitial fibroblast differentiation
response to methionine
mRNA binding
nucleoside diphosphate kinase activity
phosphoenolpyruvate carboxykinase (GTP) activity
GTP binding
inosine kinase activity
malate oxidase activity
protein domain specific binding
manganese ion binding
sequence-specific DNA binding
protein-disulfide reductase activity
DBD domain binding
epinephrine binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline