Search
Search

APOV1| Gene Symbol: APOV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205483.2 | NP_990814.2 | 3 - 4 | 325 | 93 | |
| XM_015295934.1 | XP_015151420.1 | 3 - 4 | 401 | 93 | ||
| GenBank mRNA | BU125044.1 | - | 3 - 4 | 326 | 93 | |
| BU125343.1 | - | 270 | 93 | |||
| M25774.1 | - | 3 - 4 | 308 | 93 | ||
| V00449.1 | - | 279 | 93 | |||
Set Membership: |
|