Search
Search

GLUL| Gene Symbol: GLUL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205493.1 | NP_990824.1 | 3 - 4 | 422 | 105 | |
| GenBank mRNA | M29076.1 | - | 3 - 4 | 422 | 105 | |
| S45408.1 | - | 2 - 3 | 382 | 105 | ||
Set Membership: |
|