Search
Search

TRA2A| Gene Symbol: TRA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006360.1 | NP_001006360.1 | 2 - 3 | 355 | 80 | |
| XM_025147197.1 | XP_025002965.1 | 3 - 4 | 653 | 80 | ||
| GenBank mRNA | AJ719598.1 | - | 2 - 3 | 355 | 80 | |
| BX931700.1 | - | 189 | 80 | |||
Set Membership: |
|