Search
Search

FGF8| Gene Symbol: FGF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012767.1 | NP_001012785.1 | 3 - 4 | 247 | 99 | |
| XM_015288595.2 | XP_015144081.1 | 3 - 4 | 429 | 99 | ||
| GenBank mRNA | FJ882410.1 | - | 3 - 4 | 247 | 99 | |
| U55189.1 | - | 3 - 4 | 247 | 99 | ||
Set Membership: |
|