Search
Search

YTHDF1| Gene Symbol: YTHDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012833.1 | NP_001012851.1 | 4 - 5 | 2013 | 158 | |
| GenBank mRNA | AJ719437.1 | - | 4 - 5 | 2005 | 158 | |
| AJ851651.1 | - | 4 - 5 | 2013 | 158 | ||
| CR386212.1 | - | 1 - 2 | 66 | 158 | ||
Set Membership: |
|