Search
Search

ITGA9| Gene Symbol: ITGA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012858.1 | NP_001012876.1 | 12 - 13 | 1421 | 86 | |
| GenBank mRNA | AJ719562.1 | - | 12 - 13 | 1421 | 86 | |
| BX929301.1 | - | 12 - 13 | 1164 | 86 | ||
Set Membership: |
|