Search
Search

SYNCRIP| Gene Symbol: SYNCRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031083.1 | NP_001026254.1 | 8 - 9 | 1078 | 77 | |
| XM_015284620.2 | XP_015140106.2 | 10 - 11 | 5075 | 77 | ||
| XM_015284621.2 | XP_015140107.2 | 10 - 11 | 5075 | 77 | ||
| XM_015284622.2 | XP_015140108.1 | 10 - 11 | 1421 | 77 | ||
| XM_015284623.2 | XP_015140109.2 | 10 - 11 | 5075 | 77 | ||
| XM_015284624.2 | XP_015140110.2 | 10 - 11 | 1421 | 77 | ||
| XM_025148759.1 | XP_025004527.1 | 8 - 9 | 4733 | 77 | ||
| GenBank mRNA | AJ719757.1 | - | 8 - 9 | 1101 | 77 | |
Set Membership: |
|