Search
Search

PTRF| Gene Symbol: PTRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001471.1 | NP_001001471.1 | 1 - 2 | 567 | 70 | |
| GenBank mRNA | D26315.1 | - | 1 - 2 | 593 | 70 | |
| D49739.1 | - | 1 - 2 | 567 | 70 | ||
| D82079.1 | - | 1 - 2 | 567 | 70 | ||
Set Membership: |
|