Search
Search

TENM2| Gene Symbol: TENM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003718.1 | NP_001003718.1 | 4 - 5 | 951 | 76 | |
| NM_204097.2 | NP_989428.2 | 5 - 6 | 1035 | 76 | ||
| XM_015293567.2 | XP_015149053.1 | 7 - 8 | 1308 | 76 | ||
| XM_015293568.2 | XP_015149054.1 | 7 - 8 | 1308 | 76 | ||
| XM_015293569.2 | XP_015149055.1 | 7 - 8 | 1308 | 76 | ||
| XM_015293571.2 | XP_015149057.1 | 6 - 7 | 1224 | 76 | ||
| XM_015293573.2 | XP_015149059.1 | 7 - 8 | 1308 | 76 | ||
| XM_015293576.2 | XP_015149062.1 | 7 - 8 | 1308 | 76 | ||
| XM_015293578.2 | XP_015149064.1 | 4 - 5 | 1014 | 76 | ||
| XM_015293579.2 | XP_015149065.1 | 3 - 4 | 928 | 76 | ||
| XM_015293581.2 | XP_015149067.1 | 6 - 7 | 1224 | 76 | ||
| XM_025154773.1 | XP_025010541.1 | 4 - 5 | 951 | 76 | ||
| XM_025154774.1 | XP_025010542.1 | 4 - 5 | 951 | 76 | ||
| XM_025154775.1 | XP_025010543.1 | 6 - 7 | 1098 | 76 | ||
| XM_025154776.1 | XP_025010544.1 | 4 - 5 | 951 | 76 | ||
| XM_025154777.1 | XP_025010545.1 | 5 - 6 | 1014 | 76 | ||
| XM_025154778.1 | XP_025010546.1 | 4 - 5 | 9448 | 76 | ||
| GenBank mRNA | AJ245711.1 | - | 4 - 5 | 951 | 76 | |
| AJ279031.1 | - | 5 - 6 | 1035 | 76 | ||
Set Membership: |
|