Search
Search

CYP8B1| Gene Symbol: CYP8B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005571.1 | NP_001005571.1 | 1 | 1453 | 78 | |
| XM_025147312.1 | XP_025003080.1 | 3 | 4384 | 78 | ||
| GenBank mRNA | BX934837.1 | - | 1 | 1453 | 78 | |
Set Membership: |
|