Search
Search

EPAS1| Gene Symbol: EPAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204807.2 | NP_990138.1 | 2 - 3 | 231 | 75 | |
| XM_015283618.2 | XP_015139104.1 | 2 - 3 | 975 | 75 | ||
| XM_015283619.2 | XP_015139105.1 | 2 - 3 | 975 | 75 | ||
| GenBank mRNA | AF129813.1 | - | 2 - 3 | 231 | 75 | |
Set Membership: |
|