Search
Search

TLX1| Gene Symbol: TLX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205015.1 | NP_990346.1 | 2 - 3 | 670 | 143 | |
| XM_015288580.2 | XP_015144066.2 | 2 - 3 | 1288 | 143 | ||
| GenBank mRNA | AF071874.1 | - | 2 - 3 | 670 | 143 | |
Set Membership: |
|