Search
Search

CENPC| Gene Symbol: CENPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205051.1 | NP_990382.1 | 4 - 5 | 253 | 99 | |
| XM_015276138.2 | XP_015131624.1 | 4 - 5 | 307 | 99 | ||
| GenBank mRNA | AB004649.1 | - | 4 - 5 | 253 | 99 | |
| AB004650.1 | - | 4 - 5 | 226 | 99 | ||
Set Membership: |
|