Search
Search

PAX7| Gene Symbol: PAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205065.1 | NP_990396.1 | 9 - 10 | 1402 | 102 | |
| XM_015296828.2 | XP_015152314.1 | 9 - 10 | 1566 | 102 | ||
| XM_015296829.2 | XP_015152315.1 | 9 - 10 | 1564 | 102 | ||
| XM_025142485.1 | XP_024998253.1 | 9 - 10 | 1555 | 102 | ||
| GenBank mRNA | D87838.1 | - | 9 - 10 | 1402 | 102 | |
Set Membership: |
|