Search
Search

RPL27| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205337.1 | NP_990668.1 | 2 - 3 | 104 | 77 | |
| GenBank mRNA | BX930617.1 | - | 1 - 2 | 202 | 77 | |
| X56852.1 | - | 104 | 77 | |||
Set Membership: |
|