Search
Search

MASP1| Gene Symbol: MASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213586.1 | NP_998751.1 | 4 - 5 | 702 | 81 | |
| XM_015291383.2 | XP_015146869.1 | 4 - 5 | 81 | |||
| XM_025153324.1 | XP_025009092.1 | 4 - 5 | 81 | |||
| GenBank mRNA | AY567829.1 | - | 4 - 5 | 702 | 81 | |
Set Membership: |
|