Search
Search

C11H16orf70| Gene Symbol: C11H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030565.1 | NP_001025736.1 | 7 - 8 | 530 | 80 | |
| GenBank mRNA | AJ720060.1 | - | 7 - 8 | 530 | 80 | |
Set Membership: |
|